siRNA Details
| Gene Symbol: Grk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011881.3 | NP_036011.3 | 7 | 1611 | ||
| GenBank mRNA | BC075722.1 | - | 6 | 2935 | ||
| BC096611.1 | - | 7 | 1611 | 5357325 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grk1| Gene Symbol: Grk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011881.3 | NP_036011.3 | 7 | 1611 | ||
| GenBank mRNA | BC075722.1 | - | 6 | 2935 | ||
| BC096611.1 | - | 7 | 1611 | 5357325 | ||