siRNA Details
| Gene Symbol: Rnf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037205.1 | - | 5, 6 | 498 | ||
| AF037206.1 | - | 5, 6 | 498 | |||
| AK036591.1 | - | 5, 6 | 543 | |||
| AK082296.1 | - | 5, 6 | 539 | |||
| AK139447.1 | - | 5, 6 | 504 | |||
| AK140682.1 | - | 5, 6 | 553 | |||
| AK158046.1 | - | 5, 6 | 539 | |||
| AK167278.1 | - | 5, 6 | 503 | |||
| BC058182.1 | - | 5, 6 | 506 | 5324040 | ||

Rnf13
Genomic Map