siRNA Details
| Gene Symbol: Scn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011887.3 | NP_036017.3 | 19, 20 | 3206 | ||
| GenBank mRNA | AF118044.1 | - | 19, 20 | 3206 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn11a| Gene Symbol: Scn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011887.3 | NP_036017.3 | 19, 20 | 3206 | ||
| GenBank mRNA | AF118044.1 | - | 19, 20 | 3206 | ||