siRNA Details
| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011891.4 | NP_036021.1 | 4 | 378 | ||
| GenBank mRNA | AB024923.1 | - | 4 | 599 | ||
| AK040974.1 | - | 4 | 380 | |||
| AK047384.1 | - | 1120 | ||||
| BC130019.1 | - | 4 | 368 | 40140624 | ||

Sgcd
Genomic Map