siRNA Details
| Gene Symbol: Spry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF176903.1 | - | 3 | 654 | ||
| AK014578.1 | - | 3 | 668 | |||
| AK140405.1 | - | 3 | 666 | |||
| BC039139.1 | - | 3 | 672 | 3672353 | ||
| BC051977.1 | - | 2 | 958 | |||
| BC069914.1 | - | 1 | 175 | 5251876 | ||

Spry1
Genomic Map