siRNA Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112705.2 | NP_001106176.1 | 11 | 1405 | ||
| NM_001294329.1 | NP_001281258.1 | 9 | 1265 | |||
| NM_001294330.1 | NP_001281259.1 | 9 | 1213 | |||
| NM_001294331.1 | NP_001281260.1 | 11 | 1418 | |||
| NM_001294334.1 | NP_001281263.1 | 11 | 1405 | |||
| GenBank mRNA | AF045252.1 | - | 11 | 1424 | ||
| AF045253.1 | - | 10 | 852 | |||
| AF045254.1 | - | 6 | 566 | |||
| AK014829.1 | - | 11 | 1043 | |||
| AK089925.1 | - | 9 | 793 | |||
| AK165667.1 | - | 11 | 1408 | |||
| BC066198.1 | - | 9 | 758 | |||
| CD554252.1 | - | 5 | 529 | |||
| U56909.1 | - | 2 | 103 | |||

Tlk2
Genomic Map