siRNA Details
| Gene Symbol: Tnfsf13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033622.1 | NP_296371.1 | 1 | 289 | ||
| GenBank mRNA | AF119383.1 | - | 1 | 289 | ||
| AF352245.1 | - | 1 | 293 | |||
| AK050384.1 | - | 1 | 271 | |||
| AK079180.1 | - | 1 | 450 | |||
| AK155288.1 | - | 1 | 68 | |||
| AK165081.1 | - | 1 | 280 | |||
| AY290823.1 | - | 1 | 73 | |||
| BC106840.1 | - | 1 | 262 | 40045718 | ||
| BC106841.1 | - | 1 | 262 | 40045720 | ||

Tnfsf13b
Genomic Map