siRNA Details
| Gene Symbol: Zfp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044747.2 | NP_001038212.1 | 4, 5 | 470 | ||
| NM_001163797.1 | NP_001157269.1 | 5, 6 | 560 | |||
| NM_013844.2 | NP_038872.1 | 5, 6 | 566 | |||
| GenBank mRNA | AB024005.1 | - | 5, 6 | 550 | ||
| AK013868.1 | - | 5, 6 | 561 | |||
| AK044831.1 | - | 4, 5 | 440 | |||
| AK146092.1 | - | 5, 6 | 527 | |||
| AK153887.1 | - | 5, 6 | 502 | |||
| BC018171.1 | - | 5, 6 | 549 | 3989444 | ||
| U29502.1 | - | 2, 3 | 141 | |||

Zfp68
Genomic Map