siRNA Details
| Gene Symbol: Dapp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011932.2 | NP_036062.1 | 3 | 382 | ||
| GenBank mRNA | AF163255.1 | - | 3 | 336 | ||
| AF186023.1 | - | 3 | 357 | |||
| AK145745.1 | - | 3 | 348 | |||
| AK146180.1 | - | 3 | 351 | |||
| AK169584.1 | - | 2 | 247 | |||

Dapp1
Genomic Map