siRNA Details
| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007953.2 | NP_031979.2 | 4 | 704 | ||
| GenBank mRNA | AK019222.1 | - | 4 | 665 | ||
| AK156371.1 | - | 4 | 680 | |||
| BC034165.1 | - | 4 | 647 | |||
| BC039774.1 | - | 4 | 716 | |||
| BC138586.1 | - | 4 | 664 | 8861606 | ||
| BC171958.1 | - | 4 | 664 | 9054244 | ||
| CO425338.1 | - | 3 | 354 | |||
| U85259.1 | - | 4 | 671 | |||

Esrra
Genomic Map