siRNA Details
| Gene Symbol: Fto | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011936.2 | NP_036066.2 | 3 | 462 | ||
| GenBank mRNA | AJ237917.1 | - | 3 | 448 | ||
| AK036677.1 | - | 3 | 452 | |||
| AK040866.1 | - | 3 | 440 | |||
| AK045465.1 | - | 3 | 439 | |||
| AK046935.1 | - | 3 | 421 | |||
| AK049502.1 | - | 3 | 455 | |||
| AK088881.1 | - | 3 | 448 | |||
| AK129437.1 | - | 3 | 429 | |||
| AK161060.1 | - | 3 | 421 | |||
| AK177350.1 | - | 1 | 150 | |||
| AK220146.1 | - | 1 | 13 | |||
| BC022222.1 | - | 3 | 413 | 4237261 | ||
| BC057008.1 | - | 3 | 428 | 5708558 | ||

Fto
Genomic Map