siRNA Details
| Gene Symbol: Mapkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011941.3 | NP_036071.3 | 18 | 2470 | ||
| GenBank mRNA | AB029482.1 | - | 19 | 2284 | ||
| AK122323.1 | - | 9 | 1169 | |||
| BC070449.1 | - | 18 | 2470 | 30543913 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mapkbp1| Gene Symbol: Mapkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011941.3 | NP_036071.3 | 18 | 2470 | ||
| GenBank mRNA | AB029482.1 | - | 19 | 2284 | ||
| AK122323.1 | - | 9 | 1169 | |||
| BC070449.1 | - | 18 | 2470 | 30543913 | ||