siRNA Details
| Gene Symbol: Map2k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009157.4 | NP_033183.1 | 5, 6 | 686 | ||
| GenBank mRNA | AK036928.1 | - | 5, 6 | 690 | ||
| AK044534.1 | - | 5, 6 | 678 | |||
| AK044698.1 | - | 5, 6 | 678 | |||
| AK139777.1 | - | 6, 7 | 709 | |||
| BC029833.1 | - | 3, 4 | 437 | 3962163 | ||
| U18310.1 | - | 5, 6 | 661 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001030023, AY880882, KC795688.

Map2k4
Genomic Map