siRNA Details
| Gene Symbol: Map3k11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022012.3 | NP_071295.2 | 3 | 878 | ||
| GenBank mRNA | BC047152.1 | - | 1 | 625 | 6476975 | |
| BC095963.1 | - | 3 | 878 | 5400993 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k11| Gene Symbol: Map3k11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022012.3 | NP_071295.2 | 3 | 878 | ||
| GenBank mRNA | BC047152.1 | - | 1 | 625 | 6476975 | |
| BC095963.1 | - | 3 | 878 | 5400993 | ||