siRNA Details
| Gene Symbol: Map3k12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163643.1 | NP_001157115.1 | 11 | 1868 | ||
| NM_009582.4 | NP_033608.3 | 9 | 1692 | |||
| GenBank mRNA | AK029882.1 | - | 9 | 1501 | ||
| AK034374.1 | - | 11 | 1684 | |||
| AK081623.1 | - | 4 | 360 | |||
| AK166435.1 | - | 11 | 1788 | |||
| BC047158.1 | - | 8 | 1486 | 6494216 | ||
| BC057572.1 | - | 11 | 1692 | 6401321 | ||
| U14636.1 | - | 9 | 1524 | |||
| U23789.1 | - | 8 | 1482 | |||

Map3k12
Genomic Map