siRNA Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009316.1 | NP_033342.1 | 10 | 1124 | ||
| NM_172688.3 | NP_766276.1 | 10 | 1124 | |||
| GenBank mRNA | AK046565.1 | - | 10 | 1090 | ||
| AK053911.1 | - | 10 | 1110 | |||
| AK133642.1 | - | 4 | 251 | |||
| AK137046.1 | - | 10 | 1125 | |||
| AK159277.1 | - | 10 | 1088 | |||
| BC006665.1 | - | 10 | 1115 | 3499247 | ||
| CV559615.1 | - | 3 | 180 | |||
| D76446.1 | - | 10 | 1124 | |||

Map3k7
Genomic Map