siRNA Details
| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016700.4 | NP_057909.1 | 10 | 1203 | ||
| GenBank mRNA | AB005663.1 | - | 9 | 1036 | ||
| AB206575.1 | - | 9 | 978 | |||
| AB219977.1 | - | 9 | 1012 | |||
| AK043102.1 | - | 10 | 1203 | |||
| AK139498.1 | - | 10 | 1191 | |||
| BC053027.1 | - | 10 | 1173 | 5694771 | ||

Mapk8
Genomic Map