siRNA Details
| Gene Symbol: KCNC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004977.2 | NP_004968.2 | 4 | 2528 | ||
| XM_006723203.2 | XP_006723266.1 | 4 | 2233 | |||
| XM_011526926.1 | XP_011525228.1 | 4 | 2233 | |||
| GenBank mRNA | AF055989.1 | - | 4 | 2528 | ||

KCNC3
Genomic Map