siRNA Details
| Gene Symbol: Prl2c4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011954.2 | NP_036084.2 | 809 | |||
| GenBank mRNA | BC058816.1 | - | 809 | 30053850 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl2c4| Gene Symbol: Prl2c4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011954.2 | NP_036084.2 | 809 | |||
| GenBank mRNA | BC058816.1 | - | 809 | 30053850 | ||