siRNA Details
| Gene Symbol: KCND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004979.5 | NP_004970.3 | 1 | 1861 | ||
| XM_011543910.2 | XP_011542212.1 | 2 | 769 | |||
| XM_017029508.1 | XP_016884997.1 | 2 | 600 | |||
| XM_017029509.1 | XP_016884998.1 | 2 | 597 | |||
| XM_017029510.1 | XP_016884999.1 | 1 | 1711 | |||
| GenBank mRNA | AB021865.1 | - | 1 | 1586 | ||
| AF166003.1 | - | 1 | 338 | |||
| AJ005898.1 | - | 1 | 288 | |||
| AK315092.1 | - | 1 | 772 | |||
| BC045659.1 | - | 1 | 1859 | 5271042 | ||

KCND1
Genomic Map