siRNA Details
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011958.3 | NP_036088.3 | 9 | 903 | ||
| GenBank mRNA | AJ003140.1 | - | 9 | 864 | ||
| AK010258.1 | - | 9 | 887 | |||
| AK011550.1 | - | 9 | 869 | |||
| AK049534.1 | - | 9 | 870 | |||
| AK076324.1 | - | 9 | 904 | |||
| AK084326.1 | - | 9 | 872 | |||
| AK145454.1 | - | 9 | 914 | |||
| AK169291.1 | - | 9 | 887 | |||
| BC015072.1 | - | 9 | 875 | 4212266 | ||
| BC019748.1 | - | 9 | 845 | 5007346 | ||
| Y16386.1 | - | 8 | 735 | |||

Orc4
Genomic Map