siRNA Details
| Gene Symbol: Psg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007676.4 | NP_031702.3 | 2 | 559 | ||
| GenBank mRNA | AB500066.1 | - | 1 | 135 | ||
| BC030357.1 | - | 2 | 569 | 5367680 | ||
| U34272.1 | - | 2 | 515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psg16| Gene Symbol: Psg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007676.4 | NP_031702.3 | 2 | 559 | ||
| GenBank mRNA | AB500066.1 | - | 1 | 135 | ||
| BC030357.1 | - | 2 | 569 | 5367680 | ||
| U34272.1 | - | 2 | 515 | |||