siRNA Details
| Gene Symbol: Psma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289476.1 | NP_001276405.1 | 6, 7 | 1392 | ||
| NM_011969.2 | NP_036099.1 | 6, 7 | 753 | |||
| GenBank mRNA | AF019662.1 | - | 6, 7 | 691 | ||
| AK088615.1 | - | 6, 7 | 698 | |||
| AK146765.1 | - | 6, 7 | 710 | |||
| AK165563.1 | - | 6, 7 | 706 | |||
| AK168234.1 | - | 6, 7 | 724 | |||
| BC008222.1 | - | 6, 7 | 686 | 2581897 | ||
| BC014752.1 | - | 6, 7 | 1340 | |||
| DV049235.1 | - | 6, 7 | 722 | |||

Psma7
Genomic Map