siRNA Details
Gene Symbol: Mok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011973.2 | NP_036103.1 | 3, 4 | 396 | ||
GenBank mRNA | AB022695.1 | - | 3, 4 | 396 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011973.2 | NP_036103.1 | 3, 4 | 396 | ||
GenBank mRNA | AB022695.1 | - | 3, 4 | 396 |