siRNA Details
| Gene Symbol: Rbbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015754.2 | NP_056569.2 | 5 | 469 | ||
| GenBank mRNA | AK017606.1 | - | 5 | 410 | ||
| AK076944.1 | - | 5 | 464 | |||
| BC011107.1 | - | 5 | 407 | 3982875 | ||
| BC050201.1 | - | 5 | 405 | 30013927 | ||
| BC147814.1 | - | 5 | 462 | 9087531 | ||
| BC147815.1 | - | 5 | 462 | 9087532 | ||

Rbbp9
Genomic Map