siRNA Details
| Gene Symbol: Zfp146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011980.3 | NP_036110.1 | 2 | 1018 | ||
| GenBank mRNA | AJ224763.1 | - | 2 | 981 | ||
| AK047696.1 | - | 2 | 1026 | |||
| BC127038.1 | - | 1 | 459 | 40051938 | ||
| BC127039.1 | - | 1 | 460 | 40051941 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006247118.

Zfp146
Genomic Map