siRNA Details
| Gene Symbol: Zfp146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011980.3 | NP_036110.1 | 2 | 787 | ||
| GenBank mRNA | AK047696.1 | - | 2 | 795 | ||
| BC127038.1 | - | 1 | 228 | 40051938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp146| Gene Symbol: Zfp146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011980.3 | NP_036110.1 | 2 | 787 | ||
| GenBank mRNA | AK047696.1 | - | 2 | 795 | ||
| BC127038.1 | - | 1 | 228 | 40051938 | ||