siRNA Details
| Gene Symbol: Hunk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015755.2 | NP_056570.1 | 4 | 691 | ||
| GenBank mRNA | AF055919.2 | - | 4 | 691 | ||
| AF167987.1 | - | 4 | 703 | |||
| AK144639.1 | - | 4 | 674 | |||
| AK147425.1 | - | 4 | 1076 | |||
| AK147734.1 | - | 4 | 445 | |||
| BC137788.1 | - | 4 | 752 | 8860807 | ||
| BC137790.1 | - | 4 | 751 | 8860809 | ||

Hunk
Genomic Map