siRNA Details
| Gene Symbol: Ror1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013845.4 | NP_038873.2 | 9 | 2292 | ||
| GenBank mRNA | AB010383.1 | - | 9 | 1864 | ||
| AK046699.1 | - | 9 | 2154 | |||
| AK049369.1 | - | 9 | 2293 | |||
| AK080896.1 | - | 9 | 2240 | |||
| AK087548.1 | - | 9 | 2225 | |||
| BC138794.1 | - | 9 | 2277 | 8861816 | ||
| BC138810.1 | - | 9 | 2277 | 8861832 | ||

Ror1
Genomic Map