siRNA Details
| Gene Symbol: Slc27a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011988.2 | NP_036118.2 | 2 | 828 | ||
| GenBank mRNA | AF072758.1 | - | 2 | 532 | ||
| AJ577573.1 | - | 2 | 828 | |||
| AK037042.1 | - | 2 | 588 | |||
| AK076014.1 | - | 2 | 560 | |||
| AK131156.1 | - | 1 | 1038 | |||
| AK135190.1 | - | 2 | 805 | |||
| BC060052.1 | - | 2 | 839 | |||
| BC141134.1 | - | 2 | 793 | 9055746 | ||

Slc27a3
Genomic Map