siRNA Details
| Gene Symbol: Slc7a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011990.2 | NP_036120.1 | 10 | 1576 | ||
| GenBank mRNA | AB022345.1 | - | 10 | 1543 | ||
| AK076322.1 | - | 10 | 1551 | |||
| AK089202.1 | - | 10 | 1549 | |||
| AK162233.1 | - | 10 | 1351 | |||
| AK163168.1 | - | 10 | 1550 | |||
| AK165599.1 | - | 10 | 1552 | |||
| AK171192.1 | - | 10 | 1574 | |||
| AK171252.1 | - | 10 | 1550 | |||
| AY766236.1 | - | 10 | 1543 | |||
| AY766237.1 | - | 1543 | ||||
| BC141402.1 | - | 10 | 1473 | 9056015 | ||
| BC141408.1 | - | 10 | 1473 | 9056021 | ||

Slc7a11
Genomic Map