siRNA Details
| Gene Symbol: KCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002237.3 | NP_002228.2 | 2 | 605 | ||
| XM_006723785.2 | XP_006723848.1 | 4 | 988 | |||
| XM_011528800.1 | XP_011527102.1 | 3 | 663 | |||
| XM_011528801.1 | XP_011527103.1 | 4 | 772 | |||
| XM_011528802.1 | XP_011527104.1 | 4 | 767 | |||
| XM_011528803.2 | XP_011527105.1 | 4 | 694 | |||
| XM_011528804.1 | XP_011527106.1 | 3 | 917 | |||
| XM_011528805.2 | XP_011527107.1 | 2 | 2828 | |||
| XM_011528806.1 | XP_011527108.1 | 2 | 809 | |||
| GenBank mRNA | AF033383.1 | - | 1 | 310 | ||
| AK290689.1 | - | 2 | 606 | |||
| BC006367.1 | - | 3 | 821 | 4131541 | ||
| BC046629.2 | - | 2 | 574 | 5287759 | ||
| CN301212.1 | - | 1 | 326 | |||

KCNG1
Genomic Map