siRNA Details
| Gene Symbol: Abcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011994.2 | NP_036124.2 | 1 | 885 | ||
| GenBank mRNA | AK051445.1 | - | 1 | 859 | ||
| AK082588.1 | - | 1 | 857 | |||
| AK087653.1 | - | 1 | 504 | |||
| AK161027.1 | - | 1 | 546 | |||
| BC019187.1 | - | 1 | 829 | 5027149 | ||
| Z48670.1 | - | 1 | 889 | |||

Abcd2
Genomic Map