siRNA Details
| Gene Symbol: Chst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011998.4 | NP_036128.3 | 2 | 1182 | ||
| GenBank mRNA | AF109155.1 | - | 2 | 1406 | ||
| AI614882.1 | - | 1 | 158 | |||
| AK009113.1 | - | 2 | 1183 | |||
| BC057886.1 | - | 2 | 1202 | 4987327 | ||

Chst4
Genomic Map