siRNA Details
| Gene Symbol: Clec4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170332.1 | NP_001163803.1 | 7 | 854 | ||
| NM_001170333.1 | NP_001163804.1 | 7 | 1006 | |||
| NM_011999.4 | NP_036129.1 | 6 | 934 | |||
| GenBank mRNA | AF387099.1 | - | 6 | 939 | ||
| AJ133533.1 | - | 6 | 940 | |||
| AK020363.1 | - | 4 | 405 | |||
| AK049002.1 | - | 6 | 859 | |||
| BC006623.1 | - | 7 | 855 | 3968187 | ||
| BC075729.1 | - | 6 | 776 | 30287332 | ||
| BC096565.1 | - | 7 | 863 | 874396 | ||

Clec4a2
Genomic Map