siRNA Details
| Gene Symbol: Clec4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170332.1 | NP_001163803.1 | 4 | 459 | ||
| NM_001170333.1 | NP_001163804.1 | 3 | 539 | |||
| NM_011999.4 | NP_036129.1 | 3 | 539 | |||
| GenBank mRNA | AA387662.1 | - | 3 | 545 | ||
| AF387099.1 | - | 3 | 544 | |||
| AJ133533.1 | - | 3 | 545 | |||
| AK049002.1 | - | 3 | 464 | |||
| BC006623.1 | - | 3 | 388 | 3968187 | ||
| BC075729.1 | - | 3 | 381 | 30287332 | ||
| BC096565.1 | - | 4 | 468 | 874396 | ||

Clec4a2
Genomic Map