siRNA Details
| Gene Symbol: Cops7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164089.1 | NP_001157561.1 | 6, 7 | 800 | ||
| NM_012003.2 | NP_036133.1 | 6, 7 | 785 | |||
| GenBank mRNA | AF071316.1 | - | 6, 7 | 787 | ||
| AK010417.1 | - | 6, 7 | 795 | |||
| AK010726.1 | - | 6, 7 | 770 | |||
| AK013155.1 | - | 6, 7 | 753 | |||
| AK019122.1 | - | 6, 7 | 777 | |||
| BC003724.1 | - | 5, 6 | 666 | 3593979 | ||
| CF898433.1 | - | 2, 3 | 281 | |||

Cops7a
Genomic Map