siRNA Details
| Gene Symbol: Cops7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172974.2 | NP_766562.1 | 3 | 344 | ||
| GenBank mRNA | AF071317.1 | - | 3 | 322 | ||
| AK053615.1 | - | 3 | 303 | |||
| AK081309.1 | - | 3 | 344 | |||
| AK168740.1 | - | 3 | 303 | |||
| AK170140.1 | - | 3 | 303 | |||
| BC012659.1 | - | 3 | 308 | 4017012 | ||

Cops7b
Genomic Map