siRNA Details
| Gene Symbol: Cops7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172974.2 | NP_766562.1 | 6, 7 | 792 | ||
| GenBank mRNA | AF071317.1 | - | 6, 7 | 770 | ||
| AK053615.1 | - | 5, 6 | 662 | |||
| AK081309.1 | - | 6, 7 | 792 | |||
| AK168740.1 | - | 6, 7 | 751 | |||
| AK170140.1 | - | 6, 7 | 751 | |||
| BC012659.1 | - | 6, 7 | 756 | 4017012 | ||

Cops7b
Genomic Map