siRNA Details
| Gene Symbol: Eif2s3x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012010.3 | NP_036140.1 | 7 | 694 | ||
| GenBank mRNA | AJ006587.1 | - | 7 | 684 | ||
| AK010819.1 | - | 4 | 372 | |||
| AK128949.1 | - | 7 | 693 | |||
| AK146900.1 | - | 7 | 735 | |||
| AK150095.1 | - | 7 | 701 | |||
| AK151579.1 | - | 3 | 250 | |||
| AK153568.1 | - | 3 | 250 | |||
| AK165866.1 | - | 7 | 696 | |||
| AK171317.1 | - | 7 | 695 | |||
| AK208120.1 | - | 3 | 216 | |||
| BC036993.1 | - | 7 | 692 | |||
| BC063755.1 | - | 7 | 694 | 6513251 | ||
| BC139234.1 | - | 7 | 694 | 8862256 | ||
| BC139235.1 | - | 7 | 694 | 8862257 | ||
| Gene Symbol: Eif2s3x-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076900.1 | - | 1104 | |||

Eif2s3x
Genomic Map