siRNA Details
Gene Symbol: Eif2s3y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012011.1 | NP_036141.1 | 4 | 377 | ||
GenBank mRNA | AJ006584.1 | - | 4 | 377 | ||
AK009448.1 | - | 4 | 382 | |||
AK153974.1 | - | 4 | 395 | |||
BC031450.1 | - | 4 | 373 | |||
BC043656.1 | - | 4 | 377 | 5368023 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
