siRNA Details
| Gene Symbol: H2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159513.1 | NP_001152985.1 | 3, 4 | 528 | ||
| NM_001159514.1 | NP_001152986.1 | 3, 4 | 528 | |||
| NM_001159515.1 | NP_001152987.1 | 3, 4 | 528 | |||
| NM_012015.2 | NP_036145.1 | 3, 4 | 528 | |||
| GenBank mRNA | AB071988.2 | - | 3, 4 | 464 | ||
| AF171080.1 | - | 3, 4 | 506 | |||
| AK039274.1 | - | 3, 4 | 509 | |||
| AK143498.1 | - | 3, 4 | 419 | |||
| BB633156.1 | - | 3, 4 | 509 | |||
| BC006955.1 | - | 3, 4 | 472 | 2811919 | ||

H2afy
Genomic Map