siRNA Details
| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012017.2 | NP_036147.1 | 6 | 763 | ||
| GenBank mRNA | AF083339.1 | - | 6 | 761 | ||
| BC055075.1 | - | 6 | 757 | 30056643 | ||
| CV562260.1 | - | 5 | 559 | |||
| CX564256.1 | - | 2 | 108 | |||

Zfp346
Genomic Map