siRNA Details
| Gene Symbol: Ppp2r5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081457.2 | NP_001074926.1 | 6 | 787 | ||
| NM_001081458.2 | NP_001074927.1 | 6 | 787 | |||
| NM_001135001.1 | NP_001128473.1 | 8 | 1018 | |||
| NM_012023.3 | NP_036153.2 | 6 | 787 | |||
| GenBank mRNA | AB055635.1 | - | 6 | 637 | ||
| AB055636.1 | - | 6 | 637 | |||
| AK031584.1 | - | 6 | 790 | |||
| AK129042.1 | - | 8 | 890 | |||
| BC003979.1 | - | 6 | 747 | 3489999 | ||
| U37353.1 | - | 6 | 616 | |||

Ppp2r5c
Genomic Map