siRNA Details
| Gene Symbol: Ppp2r5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081457.2 | NP_001074926.1 | 12 | 1414 | ||
| NM_001081458.2 | NP_001074927.1 | 12 | 1414 | |||
| NM_001135001.1 | NP_001128473.1 | 14 | 1645 | |||
| NM_012023.3 | NP_036153.2 | 12 | 1414 | |||
| GenBank mRNA | AB055635.1 | - | 12 | 1264 | ||
| AB055636.1 | - | 12 | 1264 | |||
| AK031584.1 | - | 12 | 1417 | |||
| AK047089.1 | - | 3 | 178 | |||
| AK129042.1 | - | 13 | 2615 | |||
| AK139212.1 | - | 1 | 1149 | |||
| AK178752.1 | - | 3 | 172 | |||
| AK196801.1 | - | 3 | 169 | |||
| AK211769.1 | - | 3 | 169 | |||
| AY389498.1 | - | 4 | 319 | |||
| BC003979.1 | - | 12 | 1374 | 3489999 | ||
| CN532075.1 | - | 4 | 364 | |||
| CX568604.1 | - | 4 | 338 | |||
| U37353.1 | - | 12 | 1243 | |||

Ppp2r5c
Genomic Map