siRNA Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012027.2 | NP_036157.2 | 13 | 1758 | ||
| NM_201245.3 | NP_957697.1 | 13 | 1758 | |||
| GenBank mRNA | AK035435.1 | - | 7 | 1048 | ||
| AK044512.1 | - | 12 | 1400 | |||
| AK154807.1 | - | 14 | 1765 | |||
| AK155630.1 | - | 13 | 1539 | |||
| AK158296.1 | - | 13 | 1747 | |||
| AK162530.1 | - | 11 | 1104 | |||
| AK170849.1 | - | 13 | 1742 | |||
| AK171089.1 | - | 13 | 1747 | |||
| BC039932.1 | - | 12 | 1282 | |||
| BC049803.1 | - | 13 | 1284 | |||
| BC063067.1 | - | 13 | 1721 | 5721396 | ||
| U73200.1 | - | 13 | 1618 | |||

Mprip
Genomic Map