siRNA Details
| Gene Symbol: KCNJ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004981.1 | NP_004972.1 | 2 | 779 | ||
| NM_152868.2 | NP_690607.1 | 2 | 941 | |||
| GenBank mRNA | BC032562.1 | - | 2 | 951 | ||
| BC039618.1 | - | 2 | 824 | |||
| BC113506.1 | - | 1 | 733 | 8322558 | ||
| BC113508.1 | - | 1 | 733 | 8322560 | ||
| CR456507.1 | - | 2 | 749 | |||
| S72503.1 | - | 2 | 780 | |||
| U07364.1 | - | 2 | 779 | |||
| U24056.1 | - | 2 | 916 | |||

KCNJ4
Genomic Map