siRNA Details
| Gene Symbol: Tpsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012034.3 | NP_036164.2 | 5 | 853 | ||
| GenBank mRNA | AF175523.1 | - | 5 | 754 | ||
| AK027982.1 | - | 4 | 679 | |||
| BC019974.1 | - | 5 | 777 | 4215589 | ||
| BC052325.1 | - | 5 | 822 | 6531021 | ||

Tpsg1
Genomic Map