siRNA Details
| Gene Symbol: Cul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012042.3 | NP_036172.1 | 22 | 2584 | ||
| GenBank mRNA | AF083216.1 | - | 21 | 2308 | ||
| AF136441.1 | - | 21 | 2433 | |||
| AF176910.1 | - | 22 | 2473 | |||
| AK147004.1 | - | 22 | 2566 | |||
| AK165978.1 | - | 22 | 2525 | |||
| AK199421.1 | - | 1 | 24 | |||
| BC029260.1 | - | 22 | 2584 | 2812400 | ||

Cul1
Genomic Map