siRNA Details
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039169.1 | NP_001034258.1 | 6 | 1168 | ||
NM_001039170.1 | NP_001034259.1 | 6 | 717 | |||
NM_023314.3 | NP_075803.2 | 6 | 717 | |||
GenBank mRNA | AF068116.1 | - | 6 | 631 | ||
AK017221.1 | - | 6 | 1169 | |||
AK018516.1 | - | 6 | 715 | |||
AK032965.1 | - | 6 | 721 | |||
AK076067.1 | - | 4 | 432 | |||
AK138055.1 | - | 6 | 656 | |||
AK150495.1 | - | 6 | 656 | |||
AK213875.1 | - | 1 | 43 | |||
BC028903.1 | - | 6 | 767 | 4166275 | ||
BC045153.1 | - | 6 | 651 | 6441327 | ||
BC049077.1 | - | 6 | 685 | 5708556 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108808, FQ219916, FQ232529, FQ233479, FQ220431, and others.
Panther Classification:
Gene Ontology Categories:
